missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress TrkB Functional Assay

Product Code. 17363209
Encompass
Change view
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17363209 1,000 dp (10 x 96-well) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17363209 Brand DiscoverX Brand No. 930463E3CP16L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress TrkB Functional Assay measures TrkB receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ eXpress TrkB Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated TrkB dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged TrkB and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry Kinase
For Use With (Application) Assay-Ready eXpress Kits
Quantity 1,000 dp (10 x 96-well)
Target Entry TrkB
Target TrkB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.