missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress Flt3 Activity Assay
Description
The PathHunter™ FLT3 Activity eXpress kit provides an easy-to-use cell based assay to study small molecule drug candidates targeting RTKs. This kit assesses RTK activation via detection of resulting FLT3 phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress a PK-tagged engineered fusion of the catalytic domain of FLT3 to the extracellular ligand binding domain of EGFR alongside an EA-tagged SH2 domain protein. Upon ligand induced receptor activation, cross-phosphorylation of the receptor leads to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | SH2 recruitment |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | Recombinant Human EGF |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | Flt3 |
| Cell Line | U2OS |
| Target | Flt3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?