missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress c-KIT Functional Assay

Product Code. 17303909
Encompass
Change view
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Product Code. Quantity unitSize
17303909 200 dp (2 x 96-well) 1 set
1 options
This item is not returnable. View return policy

Product Code. 17303909

Brand: DiscoverX 930853E3CP16M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress c-KIT Functional Assay measures c-KIT receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. eXpress kits contain all assay materials: cells, reagents, and plates. Cyno version.

The PathHunter™ eXpress c-KIT Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated c-KIT dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged c-KIT and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Cyno version of the protein.

Specifications

Assay Entry Receptor Phosphorylation
For Use With (Application) Assay-Ready eXpress Kits
Format 200 dp (2 x 96-well)
Primary Agonist Recombinant Human SCF
Quantity 200 dp (2 x 96-well)
Target Entry c-KIT
Cell Line U2OS
Target c-KIT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.