missing translation for 'onlineSavingsMsg'
Learn More

Paraxanthine, TRC

Product Code. 30007685
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007685 50 mg 50mg
17922925 100 mg 100mg
17912265 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007685 Supplier Toronto Research Chemicals Supplier No. TRCP19250050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-59-6
Molecular Formula C7 H8 N4 O2
Molecular Weight (g/mol) 180.16
Synonym 1H-Purine-2,6-dione, 3,7-dihydro-1,7-dimethyl-, Paraxanthine (6CI,7CI,8CI), 1,7-Dimethylxanthine, 3,7-Dihydro-1,7-dimethyl-1H-purine-2,6-dione, NSC 400018, 1,7-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, Caffeine Imp. F (EP), Paraxanthine
IUPAC Name 1,7-dimethyl-3H-purine-2,6-dione
SMILES CN1C(=O)Nc2ncn(C)c2C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)11(2)7(13)9-5/h3H,1-2H3,(H,9,13)
Formula Weight 180.0647
Percent Purity >95%
Chemical Name or Material Paraxanthine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.