Learn More
Paraformaldehyde-13C, TRC
Product Code. 30132847
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132847 250 mg 250mg
30133988 100 mg 100mg
30005103 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132847 Supplier Toronto Research Chemicals Supplier No. TRCP193267250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89277-65-6
Molecular Formula (13CH2O)n • H2O
Synonym Aldacide-13C, Flo-Mor-13C, Granuform-13C, Paraform-13C, TransFix-13C
SMILES O[13C]([H])(O[H])[H]
Specifications
Quantity 250 mg
Formula Weight 31.0139
Chemical Name or Material Paraformaldehyde-13C