Learn More
Paraffin, Supelco™
Product Code. 15837540
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15837540 25 kg 25kg
15824930 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15837540 Supplier Supelco Supplier No. 1071519025
 Restricted Item
Estimated Shipment: 20-11-2026
46 to 48, In block form
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8002-74-2
Specifications
Boiling Point 300°C to 500°C/1,013 hPa
Melting Point 42°C to 72°C
Product Type Tissue Embedding Media
Quantity 25 kg
Form Solid
Synonym Paraffin, Paraffin