Learn More
Tork Paper Wipes Roll W1
Shop All Tork Products
Click to view available options
Color:
White
Unit Size:
1 roll
Description
- Avoid streaky surfaces and lint deposits when cleaning glass with our Tork Wiping Paper.
- Save time when refilling dispensers, thanks to this high-capacity roll that lasts longer.
- Effectively tackle light wiping tasks and hand wiping with this multipurpose design.
- Boost efficiency - use with our Tork Floor or Wall Stand Dispensers for easy one-handed use.
Tekniske data
Tekniske data
| Product Type | Wiping Paper |
| Length (English) | 1093.61 yd. |
| Length (Metric) | 1000 m |
| Width (English) | 9.65 in. |
| Width (Metric) | 24.5 cm |
| Material | Paper |
| Certifications/Compliance | EU Ecolabel, FSC MIX Credit, FSC Chain-Of-Custody, ISO 9001, ISO 14001, ISO 45001, ISO 50001 |
| Color | White |
| Packaging Quantity | 1 Ea. |
| Ply | 1-ply |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion