Learn More
Paper Starch iodate EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA
Product Code. 12940544 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12940544 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12940544 Supplier Fisher Chemical Supplier No. RP 206 500
  • ONLINE EXCLUSIVE PRICE
    314.00 EUR
    235.53 EUR / 50mL
    Save 78.47 EUR
    24% Off
 Restricted Item
Estimated Shipment: 14-01-2027
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Analyte or Component Names Paper Starch iodate
Quantity 50 mL