Learn More
Mettler Toledo™ Paper Roll
Product Code. 12603516
Change view
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12603516 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12603516 Supplier Mettler Toledo™ Supplier No. 44000253
  • ONLINE EXCLUSIVE PRICE
    74.00 EUR
    70.30 EUR / Pack of 50
    Save 3.70 EUR
    5% Off
In Stock
Generate immediate documentation of results. Mettler Toledo™ Paper Rolls are used with dot matrix printers that can be attached to a range of balances, scales and meters. The bright white paper is 5cm wide and provides instant, clear print-outs for reliable record-keeping.
Many Mettler Toledo™ balances, scales and meters can be attached directly to small dot-matrix printers for instant documentation of results using a minimum of benchtop space. These printers use black ink cartridges and bright white rolls of normal paper that are 5cm wide.
Set of 50 paper rolls for the following dot matrix printers:
  • LCP-43
  • RS-P42
  • Sprinter
  • LCP-45
  • SQC-14
TRUSTED_SUSTAINABILITY
Specifications
Material Paper
Color White
Width (Metric) 51 mm
Finish Matte
For Use With (Equipment) Dot matrix printers: LCP-43, RS-P42, Sprinter, LCP-45 and SQC-14
Product Type Paper Roll
Quantity 50/Pk.