missing translation for 'onlineSavingsMsg'
Learn More

pan Nuclear Pore Complex Marker Antibody (39C7), Novus Biologicals™

Mouse Monoclonal Antibody

204.00 EUR - 497.00 EUR

Specifications

Antigen pan Nuclear Pore Complex Marker
Clone 39C7
Dilution Western Blot 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:500
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18233653
View Documents
Novus Biologicals
NBP1-05390-0.125ML
0.125 mL
204.00 EUR
125µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18078495
View Documents
Novus Biologicals
NBP1-05390
0.5 mL
497.00 EUR
0.50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

pan Nuclear Pore Complex Marker Monoclonal specifically detects pan Nuclear Pore Complex Marker in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

pan Nuclear Pore Complex Marker
Western Blot 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:500
Monoclonal
Supernatant
RUO
Pig, Equine, C. elegans, Yeast, Human, Mouse, Chicken, Bovine, Rat, Drosophila
Yeast nuclear preparations
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
62 kDa
39C7
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Core ESC Like Genes, Stem Cell Markers
107 kDa nucleoporin, NPC proteins, Nuclear pore complex protein Nup107, Nucleoporin 107kDa, Nucleoporin Nup107, NUP 107, NUP 84, NUP107, NUP84
IgG1
Tissue culture supernatant
Panspecific
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.