missing translation for 'onlineSavingsMsg'
Learn More

pan Nuclear Pore Complex Marker Antibody (39C7), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-05390-0.125ML

 View more versions of this product

Product Code. 18233653

  • 204.00 EUR / 125µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

pan Nuclear Pore Complex Marker Monoclonal specifically detects pan Nuclear Pore Complex Marker in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

pan Nuclear Pore Complex Marker
Monoclonal
Unconjugated
Tissue culture supernatant with 10mM Sodium Azide
Mouse
62 kDa
0.125 mL
Core ESC Like Genes, Stem Cell Markers
Panspecific
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunocytochemistry, Immunofluorescence
39C7
Western Blot 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:500
107 kDa nucleoporin, NPC proteins, Nuclear pore complex protein Nup107, Nucleoporin 107kDa, Nucleoporin Nup107, NUP 107, NUP 84, NUP107, NUP84
Yeast nuclear preparations
Tissue culture supernatant
RUO
Primary
Pig, Equine, C. elegans, Yeast, Human, Mouse, Chicken, Bovine, Rat, Drosophila
Supernatant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only