missing translation for 'onlineSavingsMsg'
Learn More

Palmitoyl-L-carnitine Hydrochloride, TRC

Product Code. 30138559
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138559 1 g 1g
17994125 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138559 Supplier Toronto Research Chemicals Supplier No. TRCP1550501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18877-64-0
Molecular Formula C23 H46 N O4 . Cl
Molecular Weight (g/mol) 436.07
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-, chloride (1:1), (2R)-, 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-, chloride, (2R)- (9CI), 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[(1-oxohexadecyl)oxy]-, chloride, (R)-, Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, palmitate, L- (8CI), Palmitic acid, ester with (3-carboxy-2-hydroxypropyl)trimethylammonium chloride, L- (8CI), (R)-3-(Palmitoyloxy)-4-(trimethylammonio)butanoate hydrochloride, Ammonium, (3-carboxy-2-hydroxypropyl)trimethyl-, chloride, palmitate, (-)-, Hexadecanoyl-L-carnitine hydrochloride, Hi-Carnitine, L-Palmitoylcarnitine chloride, Palmitoyl-L-carnitine chloride
IUPAC Name [(2R)-3-carboxy-2-hexadecanoyloxypropyl]-trimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC(=O)O[C@H](CC(=O)O)C[N+](C)(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C23H45NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)28-21(19-22(25)26)20-24(2,3)4;/h21H,5-20H2,1-4H3;1H/t21-;/m1./s1
Formula Weight 435.3115
Chemical Name or Material Palmitoyl-L-carnitine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.