accessibility menu, dialog, popup

UserName

CAS RN 18877-64-0

ClH3COO(R)OHONH3CCH3CH3

CAS RN 18877-64-0

IUPAC Name: [(2R)-3-carboxy-2-(hexadecanoyloxy)propyl]trimethylazanium chloride
Synonyms: (+)-palmitoylcarnitine chloride
Molecular Weight (g/mol): 436.07
Molecular Formula: C23H46ClNO4
InChi Key: GAMKNLFIHBMGQT-ZMBIFBSDSA-N
SMILES: [Cl-].CCCCCCCCCCCCCCCC(=O)O[C@H](CC(O)=O)C[N+](C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results