missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid Methyl Ester, 99+%, MP Biomedicals™

Product Code. 11374119 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11374119 Supplier MP Biomedicals Supplier No. 0210091001
  • ONLINE EXCLUSIVE PRICE
    78.30 EUR
    74.55 EUR / 1g
    Save 3.75 EUR
    4% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 112-39-0
Density 0.852g/mL
Assay Percent Range 99+%
Molecular Formula C17H34O2
Quantity 1 g
Synonym methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i
InChI Key FLIACVVOZYBSBS-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCC(=O)OC
IUPAC Name methyl hexadecanoate
Molecular Weight (g/mol) 270.457
PubChem CID 8181
ChEBI CHEBI:69187
Formula Weight 270.5
Physical Form Solid
Chemical Name or Material Palmitic Acid Methyl Ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.