missing translation for 'onlineSavingsMsg'
Learn More

Palladium (II) Acetate Trimer, TRC

Product Code. 30428760
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30428760 1 g
30424850 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428760 Supplier Toronto Research Chemicals Supplier No. TRCP1411001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53189-26-7
Molecular Formula C12 H18 O12 Pd3
Molecular Weight (g/mol) 673.52
Synonym Palladium(2+) Salt Acetic Acid Trimer, Palladium Acetate (Pd(OAc)2) Trimer, Acetic Acid Palladium Salt Trimer, Bis(acetato)palladium Trimer, Diacetatopalladium Trimer, Diacetoxypalladium Trimer, HyCat 1 Trimer, HyCat Base 1 Trimer, Palladium Acetate (Pd(OCOCH3)2) Trimer, Palladium Diacetate Trimer, Palladium(2+) Acetate Trimer, Palladium(2+) Diacetate Trimer, Palladium(II) Acetate Trimer, Palladous Acetate Trimer
IUPAC Name palladium(2+);hexaacetate
SMILES C#C[Pd]C#C.O=O.O=O.C#C[Pd]C#C.O=O.O=O.C#C[Pd]C#C.O=O.O=O.[HH].[HH].[HH].[HH].[HH].[HH]

Specifications

Quantity 1 g
InChI Formula InChI=1S/6C2H4O2.3Pd/c6*1-2(3)4;;;/h6*1H3,(H,3,4);;;/q;;;;;;3*+2/p-6
Formula Weight 671.7903
Chemical Name or Material Palladium (II) Acetate Trimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.