CAS RN 53189-26-7
CAS RN 53189-26-7
IUPAC Name:
hexakis(acetic acid) tripalladium
Synonyms:
hexakis(acetic acid) tripalladium
Molecular Weight (g/mol):
679.57
Molecular Formula:
C12H24O12Pd3
InChi Key:
FUKTVTHLBVPABW-UHFFFAOYSA-N
SMILES:
[Pd].[Pd].[Pd].CC(O)=O.CC(O)=O.CC(O)=O.CC(O)=O.CC(O)=O.CC(O)=O
1
–
1
of
1
results
1
Palladium (II) Acetate Trimer, TRC
CAS: 53189-26-7 Molecular Formula: C12 H18 O12 Pd3 Molecular Weight (g/mol): 673.52 Synonym: Palladium(2+) Salt Acetic Acid Trimer,Palladium Acetate (Pd(OAc)2) Trimer,Acetic Acid Palladium Salt Trimer,Bis(acetato)palladium Trimer,Diacetatopalladium Trimer,Diacetoxypalladium Trimer,HyCat 1 Trimer,HyCat Base 1 Trimer,Palladium Acetate (Pd(OCOCH3)2) Trimer,Palladium Diacetate Trimer,Palladium(2+) Acetate Trimer,Palladium(2+) Diacetate Trimer,Palladium(II) Acetate Trimer,Palladous Acetate Trimer IUPAC Name: palladium(2+);hexaacetate SMILES: C#C[Pd]C#C.O=O.O=O.C#C[Pd]C#C.O=O.O=O.C#C[Pd]C#C.O=O.O=O.[HH].[HH].[HH].[HH].[HH].[HH]