missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palbociclib isethionate, MedChemExpress
Description
Palbociclib (PD 0332991) isethionate is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib isethionate has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
Specifications
Specifications
| Chemical Name or Material | Palbociclib isethionate |
| CAS | 827022-33-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C26H35N7O6S |
| Quantity | 500 mg |
| Synonym | PD 0332991 isethionate |
| Solubility Information | H2O : 50 mg/mL (87.16 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (17.43 mM; Need ultrasonic) |
| SMILES | CC(C1=C(N2C3CCCC3)N=C(NC4=NC=C(N5CCNCC5)C=C4)N=C1)=C(C(C)=O)C2=O.OCCS(O)(=O)=O |
| Molecular Weight (g/mol) | 573.66 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?