missing translation for 'onlineSavingsMsg'
Learn More

PADI1 Antibody, Novus Biologicals™

Product Code. 30230344 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30230344 20 μL 20µL
30230115 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30230344 Supplier Novus Biologicals Supplier No. NBP33346320ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PADI1 Monoclonal antibody specifically detects PADI1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PADI1
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL
Formulation PBS (pH 7.3), 50% glycerol, 0.05% BSA
Gene Alias EC 3.5.3.15, HPAD10, hPAD-colony 10, PAD1, PDI, PDI1Protein-arginine deiminase type I, peptidyl arginine deiminase, type I, Peptidylarginine deiminase I, protein-arginine deiminase type-1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-277 of human PADI1.2).,, Sequence:, MAPKRVVQLSLKMPTHAVCVVGVEAHVDIHSDVPKGANSFRVSGSSGVEVFMVYNRTRVKEPIGKARWPLDTDADMVVSVGTASKELKDFKVRVSYFGEQEDQALGRSVLYLTGVDISLEVDTGRTGKVKRSQGDKKTWRWGPEGYGAILLVNCDRDNHRSAEPDLTHSWLMSLADLQDMSPMLLSCNGPDKLFDSHKLVLNVPFSDSKRVRVFCARGGNSLSDYKQVLGPQCLSYEVERQPGEQEIKFYVEGLTFPDADFLGLVSLSVSLVDPGTL
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29943
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.