missing translation for 'onlineSavingsMsg'
Learn More

Paclitaxel MP Biomedicals

Product Code. 11454862 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
100 mg
Unit Size:
100mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454862 1 mg 1mg
11464862 5 mg 5mg
11474862 25 mg 25mg
11484862 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11454862 Supplier MP Biomedicals Supplier No. 0219353201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 373-49-9
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00004437
InChI Key SECPZKHBENQXJG-FPLPWBNLSA-N
Synonym palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid
PubChem CID 445638
ChEBI CHEBI:28716
SMILES CCCCCC\C=C/CCCCCCCC(O)=O

Specifications

Color White
Assay Percent Range ≥98%
Quantity 1 mg
IUPAC Name (9Z)-hexadec-9-enoic acid
Formula Weight 853.9
Physical Form Powder
Chemical Name or Material Paclitaxel, From the Pacific Yew Tree
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.