accessibility menu, dialog, popup

UserName

CAS RN 373-49-9

H3COHO(Z)

CAS RN 373-49-9

IUPAC Name: (9Z)-hexadec-9-enoic acid
Synonyms: palmitoleic acid zoomaric acid oleopalmitate cis-delta(9)-hexadecenoic acid cis-delta-9-hexadecenoic acid zoomeric acid palmitolinoleic acid cis-9-palmitoleic acid oleopalmitic acid hexadecenoate
Molecular Weight (g/mol): 254.41
Molecular Formula: C16H30O2
InChi Key: SECPZKHBENQXJG-FPLPWBNLSA-N
SMILES: CCCCCC\C=C/CCCCCCCC(O)=O
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)