missing translation for 'onlineSavingsMsg'
Learn More

Paclitaxel-13C6, TRC

Product Code. 30422415
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422415 5 mg 5mg
30411971 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422415 Supplier Toronto Research Chemicals Supplier No. TRCP1325045MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C4113C6H51NO14
Molecular Weight (g/mol) 859.86
Synonym Abraxane-13C6, Genaxol-13C6, Genetaxyl-13C6, OncoGel-13C6, Onxal-13C6, Pacliex-13C6, (-)-Paclitaxel-13C6, Taxol-13C6, Yewtaxan-13C6, NSC 125973-13C6
SMILES O=C([C@@H]1OC(C)=O)[C@@]2(C)[C@H]([C@@](CO3)(OC(C)=O)[C@H]3C[C@@H]2O)[C@H](OC([13C]4=[13C]([H])[13C]([H])=[13C]([H])[13C]([H])=[13C]4[H])=O)[C@]5(O)C[C@H](OC([C@@H](O)[C@H](NC(C6=CC=CC=C6)=O)C7=CC=CC=C7)=O)C(C)=C1C5(C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C47H51NO14/c1-25-31(60-43(56)36(52)35(28-16-10-7-11-17-28)48-41(54)29-18-12-8-13-19-29)23-47(57)40(61-42(55)30-20-14-9-15-21-30)38-45(6,32(51)22-33-46(38,24-58-33)62-27(3)50)39(53)37(59-26(2)49)34(25)44(47,4)5/h7-21,31-33,35-38,40,51-52,57H,22-24H2,1-6H3,(H,48,54)/t31-,32-,33+,35+,36-,37+,38+,40-,45+,46-,47+/m0/s1/i9+1,14+1,15+1,20+1,21+1,30+1
Percent Purity >95%
Chemical Name or Material Paclitaxel-13C6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.