Learn More
Buerkle™ Packaging Buckets
Product Code. 11871101 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
5 L
10 L
Diameter (Metric)Top:
132 mm
175 mm
225 mm
268 mm
Height (Metric) With Closure:
123 mm
131 mm
132 mm
194 mm
263 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric)Top Height (Metric) With Closure Unit Size
11871101 10 L 268 mm 263 mm Each
11831091 1 L 132 mm 131 mm Each
11841091 2 L 175 mm 132 mm Each
11851091 3 L 225 mm 123 mm Each
11861091 5 L 225 mm 194 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11871101 Supplier Buerkle™ Supplier No. 23270010
  • ONLINE EXCLUSIVE PRICE
    7.87 EUR
    5.50 EUR / Pack of 1
    Save 2.37 EUR
    30% Off
In Stock
Add to basket
Edge
Add to basket
Manufactured from polypropylene for strength and durability. Buerkle™ Packaging Buckets are ideal for transport, packaging and storing of a wide range of materials.
Suitable for food or building material industries and non-aggressive chemicals in environmental protection, hosptials and household. Features strong handle and quality closure for product safety.
  • White polypropylene
  • Air-tight closure
  • Includes lid
TRUSTED_SUSTAINABILITY
Specifications
Product Type Packaging Bucket
Capacity (Metric) 10 L
Graduations None
Material Polypropylene
Autoclavable Autoclavable
Color White
Diameter (Metric)Top 268 mm
For Use With (Equipment) Suitable for The Food or Building Material Industries and Non-Aggressive Chemicals in Environmental Protection, Hospitals and The Household
Height (English) With Closure 10.35 in.
Height (Metric) With Closure 263 mm
Includes Quality Closure and Strong Lid
Quantity 1 Ea.