Learn More
VITLAB™ PA Spatula Spoons
Product Code. 15202789
Change view
Click to view available options
Length (Metric):
180 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15202789 180 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15202789 Supplier VITLAB™ Supplier No. 80596
  • ONLINE EXCLUSIVE PRICE
    40.00 EUR
    30.61 EUR / Pack of 10
    Save 9.39 EUR
    23% Off
Estimated Shipment: 22-12-2026
An excellent and economical option for manual stirring and mixing in the laboratory.
  • Glass fiber reinforced
  • Spatula tool on one end, spoon on the other
  • Easy to hold handle in the middle
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Pk.
Color Red
Product Type Spoon Spatula
Material Polyamide
Length (Metric) 180 mm