Learn More
p53 Monoclonal Antibody (DO-1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804804S
Encompass_Preferred
 View more versions of this product
Product Code. 17594503
  • 242.00 EUR / 30µL
Description
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
p53
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UMY4
Mouse
Affinity chromatography
RUO
29934
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Immunohistochemistry (Paraffin), Western Blot
DO-1
Unconjugated
SNX12
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3
Full length human recombinant protein of human SNX12 produced in E.coli.
30 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions