Learn More
p38 MAPK-IN-1, MedChemExpress
Product Code. 16475388
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475388 10 mg 10mg
16461577 50 mg 50mg
16438107 5 mg 5mg
16416006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16475388 Supplier Medchem Express Supplier No. HY12839
 Restricted Item
Estimated Shipment: 02-11-2026
MedChemExpress p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.
p38 MAPK-IN-1 (Compound 4) is a novel potent and selective inhibitor of p38 MAPK with IC50 of 68 nM. p38 MAPK-IN-1 shows sustained levels, low clearance and good bioavailability.
Specifications
Chemical Name or Material p38 MAPK-IN-1
CAS 1006378-90-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H15F2N2O
Quantity 10 mg
Solubility Information DMSO : 8.33 mg/mL (23.84 mM; Need ultrasonic)
SMILES FC1=CC(F)=C(C2=CC=NC(C2=CC=[NH+]3[O-])=C3C4=C(C)C=CC=C4)C=C1
Molecular Weight (g/mol) 349.35
Formula Weight 349.35
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease