missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
P2Y14/GPR105 Polyclonal antibody specifically detects P2Y14/GPR105 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | P2Y14/GPR105 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | G protein coupled receptor for UDP-glucose, GPR105, G-protein coupled receptor 105, KIAA0001G protein-coupled receptor 105, P2Y purinoceptor 14, P2Y(14) receptor, P2Y14, P2Y14 receptor, purinergic receptor P2Y, G-protein coupled, 14, UDP-glucose receptor |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 209-338 of human P2RY14 (NP_055694.3). ITKKIFKSHLKSSRNSTSVKKKSSRNIFSIVFVFFVCFVPYHIARIPYTKSQTEAHYSCQSKEILRYMKEFTLLLSAANVCLDPIIYFFLCQPFREILCKKLHIPLKAQNDLDISRIKRGNTTLESTDTL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?