missing translation for 'onlineSavingsMsg'
Learn More

P110δ-IN-1, MedChemExpress

Product Code. 16455626
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455626 100 mg 100mg
16497577 5 mg 5mg
16414838 10 mg 10mg
16411097 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16455626 Supplier Medchem Express Supplier No. HY114428
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress P110δ-IN-1 is a potent and selective inhibitor of P110δ extracted from patent WO 2014055647 A1, with an IC50 of 8.4 nM.

P110δ-IN-1 is a potent and selective inhibitor of P110δ extracted from patent WO 2014055647 A1, with an IC50 of 8.4 nM.

Specifications

Chemical Name or Material P110δ-IN-1
CAS 1595129-71-7
Color White
Purity Grade Notes Research
Molecular Formula C31H40N8O3S
Quantity 100 mg
Solubility Information DMSO : 4.62 mg/mL (7.64 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=NC2=CC=CC=C2N1C3=NC(N4CCOCC4)=NC(NC(C)(C)CC5=CC=CC=C5C6CCN(C)CC6)=N3)(C)=O
Molecular Weight (g/mol) 604.77
Formula Weight 604.77
Percent Purity 98.78%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.