Learn More
Velp Scientifica™ Glass Crucible P1
Product Code. 15399554 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
HU6 Hydrolysis Unit
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15399554 HU6 Hydrolysis Unit Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15399554 Supplier Velp Scientifica™ Supplier No. A00000086
Estimated Shipment: 03-12-2026
Holds samples for general laboratory purposes. Velp Scientifica™ Glass Crucible P1 is an excellent accessory for Velp Scientifica™ FIWE Series Fiber Analyzers.
  • Durable glass
  • Standardized
Specifications
For Use With (Equipment) HU6 Hydrolysis Unit
Product Type Crucible P1
Material Glass
Quantity 6 Pack