Learn More
p-Toluic-d7 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | p-Toluic Acid-d7 |
| CAS | 1219798-76-1 |
| Molecular Formula | C8 D7 H O2 |
| Quantity | 0.5 g |
| Synonym | Benzoic-2,3,5,6-d4 acid, 4-(methyl-d3)- (ACI), 4-(Methyl-d3)benzoic-2,3,5,6-d4 acid (ACI), p-Toluic-α,α,α,2,3,5,6-d7 acid, 4-Methylbenzoic acid D7, p-Toluic acid-d7 |
| InChI Formula | InChI=1 S/C8H8O2/c1-6-2-4-7(5-3-6)8(9)10/h2-5 H,1H3,(H,9,10)/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-(trideuteriomethyl)benzoic acid |
| Molecular Weight (g/mol) | 143.191 |
| Formula Weight | 143.096 g/mol |
| Show More |