missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic Acid Hydrate, TRC

Product Code. 30456457
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456457 10 mg 10mg
30451083 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456457 Supplier Toronto Research Chemicals Supplier No. TRCT73386410MG
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Toluenesulfonic Acid Hydrate
Molecular Formula C7H8O3S • x(H2O)
Quantity 10 mg
Synonym 4-Methylbenzenesulfonic Acid Hydrate, 4-Toluenesulfonic Acid Hydrate, Tosic Acid Hydrate, PTSA Hydrate, p-Toluenesulfonic Acid Hydrate, TsOH, p-tsa, Lisinopril EP Impurity B, Anastrozole EP Impurity F
InChI Formula InChI=1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2
SMILES O=S(C1=CC=C(C)C=C1)(O)=O.O
IUPAC Name 4-methylbenzenesulfonic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.