missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl-β-D-galactopyranoside, >99.75%, MP Biomedicals™

Product Code. 11483740 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11483740

Brand: MP Biomedicals 0210247205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 3150-24-1
Color White
Assay Percent Range > 99.75%
Molecular Formula C12H15NO8
MDL Number MFCD00063256
Quantity 5 g
Synonym 4-nitrophenyl-beta-d-galactopyranoside, 4-nitrophenylgalactoside, p-nitrophenyl-beta-d-galactoside, p-nitrophenyl beta-d-galactopyranoside, 4-nitrophenyl beta-d-galactopyranoside, p-nitrophenol galactoside, 4-nitrophenyl-beta-d-galactoside, 4-nitrophenyl galactoside, 1-o-p-nitrophenyl-beta-d-galactopyranose, 4-nitrophenylbeta-d-galactopyranoside
InChI Key IFBHRQDFSNCLOZ-YBXAARCKSA-N
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol) 301.25
PubChem CID 65115
ChEBI CHEBI:355715
Formula Weight 301.3
Physical Form Crystalline
Chemical Name or Material p-Nitrophenyl-β-D-galactopyranoside, For TLC analysis
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.