missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl Acetate, MP Biomedicals™

Product Code. 11304099 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304099 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11304099 Supplier MP Biomedicals Supplier No. 0210088805
  • ONLINE EXCLUSIVE PRICE
    49.60 EUR
    47.25 EUR / 5g
    Save 2.35 EUR
    4% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 830-03-5
Color White to Yellow
Molecular Formula C8H7NO4
MDL Number MFCD00007326
Quantity 5 g
Synonym p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester
InChI Key QAUUDNIGJSLPSX-UHFFFAOYSA-N
SMILES CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
IUPAC Name 4-nitrophenyl acetate
Molecular Weight (g/mol) 181.15
PubChem CID 13243
ChEBI CHEBI:82635
Formula Weight 181.1
Physical Form Crystalline Powder
Chemical Name or Material p-Nitrophenyl acetate
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.