Learn More
p-Nitrophenyl 2-acetamido-2-deoxy-a-D-glucopyranoside, MP Biomedicals
Product Code. 11449191 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449191 100 mg 100mg
11429191 10 mg 10mg
11439191 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11449191 Supplier MP Biomedicals Supplier No. 0215590180
  • ONLINE EXCLUSIVE PRICE
    921.00 EUR
    876.75 EUR / 100mg
    Save 44.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Chemical Identifiers
CAS 10139-02-3
Molecular Formula C14H18N2O8
Molecular Weight (g/mol) 342.304
InChI Key OMRLTNCLYHKQCK-KSTCHIGDSA-N
Synonym 4'-nitrophenyl-2-acetamido-2-deoxy-alpha-d-glucopyranoside, p-nitrophenyl n-acetyl-alpha-d-glucosaminide, n-2r,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-a-d-glucopyranoside, 4-nitrophenyl 2-acetamido-2-deoxy-alpha-d-glucopyranoside, n-2r,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-, a-d-glucopyranoside, 4-nitrophenyl n-acetyl-, a-d-glucosaminide, p-nitrophenyl-n-acetyl-alpha-d-glucosaminide, p-nitrophenyl 2-acetamido-2-deoxy-
PubChem CID 82398
IUPAC Name N-[(2R,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1OC2=CC=C(C=C2)[N+](=O)[O-])CO)O)O