missing translation for 'onlineSavingsMsg'
Learn More

p-Nitro blue tetrazolium chloride, MP Biomedicals™

Product Code. 11443090 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
0.25 g
0.5 g
1 g
5 g
Unit Size:
1g
250mg
500mg
50mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443090 5 g 5g
11403090 50 mg 50mg
11413090 0.25 g 250mg
11423090 0.5 g 500mg
11433090 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11443090 Supplier MP Biomedicals Supplier No. 0210041694
  • ONLINE EXCLUSIVE PRICE
    1045.00 EUR
    995.40 EUR / 5g
    Save 49.60 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 298-83-9
Molecular Formula C40H30Cl2N10O6
Molecular Weight (g/mol) 817.644
MDL Number MFCD00012159
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
Synonym 3,3'-(3,3'-Dimethoxy-4,4'-biphenylene)-bis-(2-p-nitrophenyl)-5-(phenyl)-2H-tetrazolium chloride, NBT
PubChem CID 9281
ChEBI CHEBI:9505
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]

Specifications

Assay Percent Range 0.981
Formula Weight 817.6
Color Yellow
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material p-Nitro blue tetrazolium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.