missing translation for 'onlineSavingsMsg'
Learn More

p-Butyl Ibuprofen, TRC

Product Code. 30011707
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011707 2 mg 2mg
17923665 1 mg 1mg
17940745 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011707 Supplier Toronto Research Chemicals Supplier No. TRCB6263052MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3585-49-7
Molecular Formula C13 H18 O2
Molecular Weight (g/mol) 206.28
Synonym (2RS)-2-(4-Butylphenyl)propanoic acid, Ibuprofen Imp. B (EP), Benzeneacetic acid, 4-butyl-α-methyl-, Hydratropic acid, p-butyl- (7CI,8CI), 4-Butyl-α-methylbenzeneacetic acid, 2-(4-Butylphenyl)propanoic acid, 2-(4-n-Butylphenyl)propionic acid, 2-(p-Butylphenyl)propionic acid, p-Butylhydratropic acid, p-Butyl Ibuprofen
IUPAC Name 2-(4-butylphenyl)propanoic acid
SMILES CCCCc1ccc(cc1)C(C)C(=O)O

Specifications

Quantity 2 mg
InChI Formula InChI=1S/C13H18O2/c1-3-4-5-11-6-8-12(9-7-11)10(2)13(14)15/h6-10H,3-5H2,1-2H3,(H,14,15)
Formula Weight 206.1307
Percent Purity >95%
Chemical Name or Material (2RS)-2-(4-Butylphenyl)propanoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.