missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Butyl Ibuprofen (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-(4-Butylphenyl)propanoic Acid |
| CAS | 3585-49-7 |
| Molecular Formula | C13 H18 O2 |
| Quantity | 100 mg |
| Synonym | (2RS)-2-(4-Butylphenyl)propanoic acid, Ibuprofen Imp. B (EP), Benzeneacetic acid, 4-butyl-α-methyl-, Hydratropic acid, p-butyl- (7CI,8CI), 4-Butyl-α-methylbenzeneacetic acid, 2-(4-Butylphenyl)propanoic acid, 2-(4-n-Butylphenyl)propionic acid, 2-(p-Butylphenyl)propionic acid, p-Butylhydratropic acid, p-Butyl Ibuprofen |
| InChI Formula | InChI=1S/C13H18O2/c1-3-4-5-11-6-8-12(9-7-11)10(2)13(14)15/h6-10H,3-5H2,1-2H3,(H,14,15) |
| SMILES | CCCCc1ccc(cc1)C(C)C(=O)O |
| IUPAC Name | 2-(4-butylphenyl)propanoic acid |
| Molecular Weight (g/mol) | 206.28 |
| Formula Weight | 206.1307 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?