missing translation for 'onlineSavingsMsg'
Learn More

p-Bromophenol, 99.5%, MP Biomedicals™

Product Code. 11419041 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419041 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11419041 Supplier MP Biomedicals Supplier No. 0215489083

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 106-41-2
Color White
Assay Percent Range 0.995
Molecular Formula C6H5BrO
Quantity 250 g
Synonym p-bromophenol, phenol, 4-bromo, p-bromohydroxybenzene, 4-bromo-phenol, p-bromophenic acid, phenol, p-bromo, para-bromophenol, 4-bromo phenol, unii-lao4j0183i, ccris 632
InChI Key GZFGOTFRPZRKDS-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1O)Br
IUPAC Name 4-bromophenol
Molecular Weight (g/mol) 173.009
PubChem CID 7808
Formula Weight 173.01
Physical Form Crystalline Powder
Chemical Name or Material p-Bromophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.