Learn More
p-Aminophenyl Beta-D-Cellobioside, TRC
Specifications
Specifications
| Chemical Name or Material | p-Aminophenyl beta-D-Cellobioside |
| CAS | 42935-24-0 |
| Molecular Formula | C18 H27 N O11 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C18H27NO11/c19-7-1-3-8(4-2-7)27-17-15(26)13(24)16(10(6-21)29-17)30-18-14(25)12(23)11(22)9(5-20)28-18/h1-4,9-18,20-26H,5-6,19H2/t9-,10-,11-,12+,13-,14-,15-,16-,17-,18+/m1/s1 |
| SMILES | Nc1ccc(O[C@@H]2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)cc1 |
| IUPAC Name | (2S,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-(4-aminophenoxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 433.41 |
| Formula Weight | 433.1584 |