Learn More
p-Amino-L-phenylalanine, TRC
Product Code. 30179544
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179544 25 g 25g
30179189 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30179544 Supplier Toronto Research Chemicals Supplier No. TRCA62237625G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 943-80-6
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.2
Synonym 4-Amino-L-phenylalanine, L-3-(p-Aminophenyl)alanine, 4-Amino-L-phenylalanine, 4-Aminophenylalanine, Aminophenylalanine, L-(+)-p-Aminophenylalanine, L-4-Aminophenylalanine;
IUPAC Name (2S)-2-amino-3-(4-aminophenyl)propanoic acid
SMILES N[C@@H](Cc1ccc(N)cc1)C(=O)O
Specifications
InChI Formula InChI=1S/C9H12N2O2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,10-11H2,(H,12,13)/t8-/m0/s1
Quantity 25 g
Formula Weight 180.0899
Shipping Condition Room Temperature
Chemical Name or Material p-Amino-L-phenylalanine