Learn More
Oxygen Cylinder Trolleys
Product Code. 12925961
Encompass
Change view
Click to view available options
Dimensions (L x W x H):
420 x 400 x 1100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
12925961 420 x 400 x 1100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12925961 Supplier Brand Not Defined Supplier No. TS178L
This item requires a price update prior to being ordered.
Request a Price for this item here.
The tubular frame has a sheet steel base and retaining straps to hold the cylinder in position. Each trolley is fitted with anti-static metal centred wheels and front 100mm non-marking corner buffers.
Specifications
Height (Metric) 1100 mm
Dimensions (L x W x H) 420 x 400 x 1100 mm
Length (Metric) 420 mm
Width (Metric) 400 mm
Holds Oxygen Cylinder
Product Type Oxygen Cylinder Trolleys
For Use With (Equipment) Oxygen Cylinder