missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WL Nutrient Broth (Dehydrated)

Product Code. 11973962
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11973962 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11973962 Supplier Thermo Scientific™ Supplier No. CM0501B

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Determine the flora in brewing and fermentation processes with this selective medium.

Determine the flora in brewing and fermentation processes with Thermo Scientific™ Oxoid™ WL Nutrient Broth (Dehydrated). The broth is based on the formulation of Green and Gray1 and is used where there are advantages for broth media e.g. using larger samples of liquid products or for enrichment cultures with cycloheximide.

Use WL Nutrient Broth for determination of the flora in brewing and fermentation processes.

  • Selective: the presence of cycloheximide in the medium suppresses the yeast growth and renders the medium selective for bacterial contaminants
  • Baker’s yeast counts can be carried out in this medium at a pH 5.5, while at pH 6.5, the medium can be used for obtaining counts of Baker’s and distiller’s yeast.

Not all products are available for sale in all territories. Please inquire.

TRUSTED_SUSTAINABILITY

Specifications

Description WL Nutrient Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 8.3L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.