Learn More
Thermo Scientific™ Oxoid™ Tryptone
Product Code. 10301783
Change view
Click to view available options
Quantity:
500 g
5 kg
25 kg
Unit Size:
2.5kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10301783 5 kg 5kg
10158962 500 g 500g
15852387 25 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10301783 Supplier Thermo Scientific™ Supplier No. LP0042T
  • ONLINE EXCLUSIVE PRICE
    1503.00 EUR
    1226.96 EUR / 5kg
    Save 276.04 EUR
    18% Off
 Restricted Item
Estimated Shipment: 20-11-2026
The Thermo Scientific™ Oxoid Tryptone is a high performance pancreatic digest of casein.
The Thermo Scientific™ Oxoid Tryptone ia a pancreatic digest of casein for use where high performance and uniformity of composition are of paramount importance.
The Thermo Scientific™ Oxoid Tryptone ia a pancreatic digest of casein for use where high performance and uniformity of composition are of paramount importance.
With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance and scale to ensure successful microbial vaccine production.
Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.
Supports quality media design:
  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free
Multi-method suitability:
  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Dehydrated Powder, Ambient & Dry Storage
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Quantity 5 kg
Protein Source Casein
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.