Learn More
Thermo Scientific™ Oxoid™ Peptone Mycological
Product Code. 10630195
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10630195 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10630195 Supplier Thermo Scientific™ Supplier No. LP0040B
  • ONLINE EXCLUSIVE PRICE
    438.14 EUR
    408.20 EUR / 500g
    Save 29.94 EUR
    6% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Oxoid Peptone Mycological is a mycological peptone developed for isolation of pathogenic and nonpathogenic fungi.
TRUSTED_SUSTAINABILITY
Specifications
Description Peptone Mycological
Form Powder
Product Type Peptone
Quantity 500 g