Learn More
Thermo Scientific™ Oxoid™ Legionella GVPC Selective Supplement
Product Code. 17100671
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17100671 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17100671 Supplier Thermo Scientific™ Supplier No. SR0252E
  • ONLINE EXCLUSIVE PRICE
    343.00 EUR
    305.46 EUR / Pack of 10
    Save 37.54 EUR
    10% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Oxoid Legionella GVPC Selective Supplement is used for the isolation of Legionella spp. from environmental samples.
Oxoid Legionella GVPC Selective Supplement is used for the isolation of Legionella spp. from environmental samples.
  • Add to Legionella Agar Base, Part No. CM1203B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Legionella GVPC Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Legionella
compliance-icons compliance-icons
Product Identifier
  • Legionella Selective Supplement (GVPC)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H301-Toxic if swallowed.
  • H341-Suspected of causing genetic defects.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Cycloheximide, Polymyxin B sulfate