Learn More
Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial
Product Code. 10065732
Change view
Click to view available options
Format:
Dip Slide
Media Type:
MacConkey Agar # 3
MacConkey Agar # 3 + germicide
OGYE Agar
Plate Count Agar
TSA (Caso) / Malt Extr + CAM
TTC (Red Spot)
TTC (Red Spot)/Malt Ext Agar
VRBG Agar + germicide
VRGB Agar
Manufacturing Origin:
Italy
United Kingdom
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Media Type Manufacturing Origin unitSize
10065732 Dip Slide Plate Count Agar Italy Pack of 10
11443959 Dip Slide MacConkey Agar # 3 United Kingdom Pack of 10
11453959 Dip Slide MacConkey Agar # 3 + germicide United Kingdom Pack of 10
11483959 Dip Slide OGYE Agar United Kingdom Pack of 10
11463959 Dip Slide VRGB Agar United Kingdom Pack of 10
11473959 Dip Slide VRBG Agar + germicide United Kingdom Pack of 10
10147942 Dip Slide TSA (Caso) / Malt Extr + CAM Italy Pack of 10
11423959 Dip Slide TTC (Red Spot) United Kingdom Pack of 10
11433959 Dip Slide TTC (Red Spot)/Malt Ext Agar United Kingdom Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10065732 Supplier Thermo Scientific™ Supplier No. GFD07A
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial.
Make colony counting simple for aerobic bacteria, yeast, mold, coliform bacteria and Enterobacteriaceae with the easily disposable Thermo Scientific™ Oxoid™ Dip-Slides™ Industrial. Simple-to-use for the operator and where laboratory facilities are limited, the Dip Slide is coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective.
Originally designed to determine bacterial counts on urine specimens, dip slides have been modified over the years by making the device double-sided, with one side a medium for Gram positive species and the other side a medium for Gram negatives. With these dip slides, the process for colony counting is time- and cost-effective, allowing exposure of the specimens to two different media simultaneously.
  • Slide is made of easily disposable plastic with raised edges to ensure an even thickness of culture medium
  • Molded grid allows for easier colony counting
  • Slide is shaped to ensure drainage of fluid from the slide
  • Cap forms a convenient handle by which it may be held without risk of touching the culture medium
  • Bacteria, fungal and yeast charts are included, make examining growth simple for the operator and where laboratory facilities are limited
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 15°C to 25°C
Format Dip Slide
Manufacturing Origin Italy
Media Type Plate Count Agar
Product Type Dip Slide
Quantity 10/Pk.