Learn More
Thermo Scientific™ Oxoid™ Clostridium Difficile Selective Supplement
Product Code. 11484169
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484169 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11484169 Supplier Thermo Scientific™ Supplier No. SR0096E
  • ONLINE EXCLUSIVE PRICE
    218.00 EUR
    210.94 EUR / Pack of 10
    Save 7.06 EUR
    3% Off
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Oxoid Clostridium Difficile Selective Supplement is added to Clostridium Difficile Agar Base, Part No. CM0601B.
Oxoid Clostridium Difficile Selective Supplement is added to Clostridium Difficile Agar Base, Part No. CM0601B.
Each vial supplements 500mL of medium.
TRUSTED_SUSTAINABILITY
Specifications
Description Clostridium Difficile Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
compliance-icons
Product Identifier
  • Clostridrium difficile SELECTIVE SUPPLEMENT
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Cefoxitin (FOX)