Learn More
Thermo Scientific™ Oxoid™ CampyGen™ Compact Sachet
Product Code. 10689374
Change view
Click to view available options
Quantity:
20/Pk.
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10689374 20/Pk. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10689374 Supplier Thermo Scientific™ Supplier No. CN0020C
  • ONLINE EXCLUSIVE PRICE
    60.10 EUR
    52.05 EUR / Pack of 20
    Save 8.05 EUR
    13% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen Compact Sachet.
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen Compact Sachet.
  • For use in Compact Plastic Pouch
  • Each pouch holds 2 standard size petri dishes
  • No water or catalyst required
  • No hydrogen gas produced
  • Use with Compact Plastic Pouch Part No. AG0020C and AnaeroGen™ Compact Sealing Clip, Part No. AN0005C
TRUSTED_SUSTAINABILITY
Specifications
Type CampyGen Compact Sachet
Format Compact Gas Generation Sachet
Product Line Thermo Scientific Oxoid
Quantity 20/Pk.