Learn More
Thermo Scientific™ Oxoid™ AnaeroJar™ Clip
Product Code. 10679754
Change view
Click to view available options
Quantity:
2/Pk.
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10679754 2/Pk. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10679754 Supplier Thermo Scientific™ Supplier No. AG0031A
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge
Add to basket
Oxoid AnaeroJar Clip has two self-venting clips.
Oxoid AnaeroJar Clip has two self-venting clips.
For use with AnaeroJar 2.5L, Part No. AG0025A
Includes:
2 self-venting clips.
TRUSTED_SUSTAINABILITY
Specifications
Type AnaeroJar Clip
Format Self-venting Clip
Includes 2 self-venting clips
Product Line Thermo Scientific Oxoid
Quantity 2/Pk.