Learn More
Thermo Scientific™ Oxoid™ Agar Bacteriological
Product Code. 10351303
Change view
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10351303 500 g 500g
10055782 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10351303 Supplier Thermo Scientific™ Supplier No. LP0011B
Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.
Oxoid Agar Bacteriological (Agar No. 1) is a clear purified high gel strength bacteriological agar compatible with all culture media.
TRUSTED_SUSTAINABILITY
Specifications
Description Agar No. 1, bacteriological
Form Powder
Product Type Agar
Quantity 500 g