Learn More
Diversey™ Oxivir Excel Concentrate
Product Code. 17231614
Change view
Click to view available options
Description:
For AT, CH, DE, DK, FI, FR, GB, IE, NO, SE only
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
17231614 For AT, CH, DE, DK, FI, FR, GB, IE, NO, SE only Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17231614 Supplier Diversey™ Supplier No. 100941436
  • ONLINE EXCLUSIVE PRICE
    205.70 EUR
    180.15 EUR / Pack of 2
    Save 25.55 EUR
    12% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Highly concentrated one step cleaner disinfectant for non-invasive medical devices and nonporous surfaces.
  • Effective against Bacteria, Viruses and Yeasts
  • Full virucidal claim including non-enveloped and enveloped viruses such as SARS-CoV-2 in just 30 sec
  • Extensively tested against European norms
  • Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
  • Excellent user safety profile reduces worker risk
  • No hazard warnings in dilution
  • Based on Diversey's patented AHP formulation (Accelerated Hydrogen Peroxide)
  • AHP formulation is gentle on surfaces extending equipment life
  • Does not contain respiratory irritants
  • Medical Device Class IIa
  • Oxivir Excel can be used on any hard surface usually encountered in a laboratory, production and non-production environments including stainless steel, glass, acrylics and PVC. It is not suitable for use on marble, copper, brass or other surfaces sensitive to oxidative agents.
Specifications
Product Type Disinfectant Concentrate
pH 1.95
Sterility Non-sterile
Composition Patented Accelerated Hydrogen Peroxide formulation (conc.)
Container Type Jug
Description For AT, CH, DE, DK, FI, FR, GB, IE, NO, SE only
Quantity 2 Pk.
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • Oxivir Excel
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
Supplemental information
  • MIXTURE LIST-Contain: alkylbenzenesulphonic acid, propanol, hydrogen peroxide
The surfactants used in this product are biodegradable in compliance with the requirement of EU Detergent Regulation, EC 648/2004